Learn More
Chemglass Life Sciences 250mL Glassblowers Flat Bottom Flask Blank
SDP
Catalog No. 501531804
Encompass_Preferred
250mL Glassblowers Flat Bottom Flask Blank. Blanks for fabrication of single or multi-neck flat bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-153-1804 Supplier Chemglass Life Sciences Supplier No. CG61903
250mL Glassblowers Flat Bottom Flask Blank. Blanks for fabrication of single or multi-neck flat bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.