Learn More
Wilmad Labglass FLASK CALB/MEAS TC 750ML
Catalog No. 50801874
Encompass
Flask, Calibration/Measuring, To Contain, Metric Series. Nominal Capacity: 750mL; Subdivision: 1.00mL. Flask is calibrated to contain at 68degreeF/20degreeC.
Catalog No. 50-801-874 Supplier Wilmad Labglass Supplier No. LG4319112
Flask, Calibration/Measuring, To Contain, Metric Series. Nominal Capacity: 750mL; Subdivision: 1.00mL. Flask is calibrated to contain at 68degreeF/20degreeC.