Learn More
Bellco Glass, Inc. Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Plug Seal Flask Size Fits 182cm2 amp 600mL Case Quantity 5
SDP
Catalog No. 502329742
Encompass_Preferred
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Plug Seal Flask Size Fits 182cm2 amp 600mL Case Quantity 5
Catalog No. 50-232-9742 Supplier Bellco Glass, Inc. Supplier No. 229398000
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Plug Seal Flask Size Fits 182cm2 amp 600mL Case Quantity 5