Learn More
Bellco Glass, Inc. Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Plug Seal Flask Size Fits 12.5cm and 25mL Case Quantity 5
SDP
Catalog No. 502330517
Encompass_Preferred
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Plug Seal Flask Size Fits 12.5cm and 25mL Case Quantity 5
Catalog No. 50-233-0517 Supplier Bellco Glass, Inc. Supplier No. 229392000
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Plug Seal Flask Size Fits 12.5cm and 25mL Case Quantity 5