Learn More
Bellco Glass, Inc. Flask Cap, Vent (fits 12.5cm2
SDP
Catalog No. 502330581
Encompass_Preferred
DescriptionReplacement caps available for Tissue Culture FlasksVent caps feature 0.22µm hydrophobic membrane Pack Description5/BagCase Quantity5SterileYesNon-pyrogenicYesPrimary ColorGreenPrimary MaterialHDPEPore Size0.22μmRNase/DNase FreeYes
Catalog No. 50-233-0581 Supplier Bellco Glass, Inc. Supplier No. 229393000
DescriptionReplacement caps available for Tissue Culture FlasksVent caps feature 0.22µm hydrophobic membrane Pack Description5/BagCase Quantity5SterileYesNon-pyrogenicYesPrimary ColorGreenPrimary MaterialHDPEPore Size0.22μmRNase/DNase FreeYes