Learn More
Bellco Glass, Inc. Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Vent Flask Size Fits 182cm 300cm2 amp 600mL amp 850mL Case Quantity 5
SDP
Catalog No. 502330093
Encompass_Preferred
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Vent Flask Size Fits 182cm 300cm2 amp 600mL amp 850mL Case Quantity 5
Catalog No. 50-233-0093 Supplier Bellco Glass, Inc. Supplier No. 229399000
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Vent Flask Size Fits 182cm 300cm2 amp 600mL amp 850mL Case Quantity 5