Learn More
Bellco Glass, Inc. Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Vent Flask Size Fits 75cm2 amp 250mL Case Quantity 5
SDP
Catalog No. 502330548
Encompass_Preferred
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Vent Flask Size Fits 75cm2 amp 250mL Case Quantity 5
Catalog No. 50-233-0548 Supplier Bellco Glass, Inc. Supplier No. 229397000
Replacement caps available for Tissue Culture Flasks. Vent caps feature 0.22¿m hydrophobic membrane.Specifications Cap Style Vent Flask Size Fits 75cm2 amp 250mL Case Quantity 5