Learn More
Quality Biological Inc Vent Cap for Tissue Culture and Suspension Culture Flasks, Small (fits 12.5 & 25 cm², 25 & 50mL), 0.22 µm hydrophobic membrane, Sterile, 5/CS
SDP
Catalog No. 50147437
Encompass_Preferred
Flask Cap, Vent (fits 12.5 cm² & 25 mL), Sterile, 5/CS
Replacement caps available for Tissue Culture Flasks
Vent caps feature 0.22µm hydrophobic membrane
Catalog No. 50-147-437 Supplier Quality Biological Inc Supplier No. P886229393
Flask Cap, Vent (fits 12.5 cm² & 25 mL), Sterile, 5/CS
Replacement caps available for Tissue Culture Flasks
Vent caps feature 0.22µm hydrophobic membrane