Learn More
Bellco Glass, Inc. Flask Carrier, 15 L
SDP
Catalog No. 501945984
Encompass_Preferred
Stainless steel bottom base for extra strength includes four rubber feet for added stability. Carriers are dishwasher and oven safe. For use with 6L or 9L vessels or carboys. MADE IN THE USA Material: stainless steel Dimensions: 2x11" Size: 15l
Catalog No. 50-194-5984 Supplier Bellco Glass, Inc. Supplier No. 766915000
Stainless steel bottom base for extra strength includes four rubber feet for added stability. Carriers are dishwasher and oven safe. For use with 6L or 9L vessels or carboys. MADE IN THE USA Material: stainless steel Dimensions: 2x11" Size: 15l