Learn More
Supply Solutions Benchmark Scientific - Best Value! Magic Clamp Magnetic Clamp, 1000Ml Erlenmeyer (Max. 5); Capacity (ML): 1000
Catalog No. NC1087832
Encompass
Accessories for Platforms H1000-MR and BT3000-MR, these flask clamps for Erlenmeyer flask are magnetic and attach to both of these platforms.; Capacity (ML): 1000
Catalog No. NC1087832 Supplier Supply Solutions Supplier No. BENCHMARK H1000MR1000
Accessories for Platforms H1000-MR and BT3000-MR, these flask clamps for Erlenmeyer flask are magnetic and attach to both of these platforms.; Capacity (ML): 1000