Learn More
Bellco Glass, Inc. Flask Clamp, 125mL
SDP
Catalog No. 501946152
Encompass_Preferred
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 9 51x51cm tray accepts 25
Catalog No. 50-194-6152 Supplier Bellco Glass, Inc. Supplier No. 774416125
Add to Cart
Add to Cart
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 9 51x51cm tray accepts 25