Learn More
Bellco Glass, Inc. Flask Clamp, 25 mL
SDP
Catalog No. 501946149
Encompass_Preferred
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 16 51x51cm tray accepts 49
Catalog No. 50-194-6149 Supplier Bellco Glass, Inc. Supplier No. 774416025
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 16 51x51cm tray accepts 49