Learn More
Bellco Glass, Inc. Flask Clamp, 250mL
SDP
Catalog No. 501946154
Encompass_Preferred
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 5 51x51cm tray accepts 16
Catalog No. 50-194-6154 Supplier Bellco Glass, Inc. Supplier No. 774416250
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 5 51x51cm tray accepts 16