Learn More
Bellco Glass, Inc. Flask Clamp, 500mL
SDP
Catalog No. 501946155
Encompass_Preferred
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 4 51x51cm tray accepts 9
Catalog No. 50-194-6155 Supplier Bellco Glass, Inc. Supplier No. 774416500
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 4 51x51cm tray accepts 9