Learn More
Bellco Glass, Inc. Flask Clamp, 50mL
SDP
Catalog No. 501946150
Encompass_Preferred
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 16 51x51cm tray accepts 49
Catalog No. 50-194-6150 Supplier Bellco Glass, Inc. Supplier No. 774416050
Flask clamps and universal platforms for shakers. Fits onto standard platforms. Purchase individual pieces to build various configurations.. MADE IN THE USA 25x25cm tray accepts 16 51x51cm tray accepts 49