Learn More
Ace Glass, Inc. Flask, 3000mL, cylindrical, flat bottom, 168mm OD x 130mm flat flange
SDP
Catalog No. 50973120
Encompass_Preferred
Flask, 3000mL, cylindrical, flat bottom, 168mm OD x 130mm flat flange
Catalog No. 50-973-120 Supplier Ace Glass, Inc. Supplier No. 651129
Flask, 3000mL, cylindrical, flat bottom, 168mm OD x 130mm flat flange