Learn More
Chemglass Life Sciences Flask, Dewar, 1000mL, Narrow Mouth, Stainless Steel, Carrying Handle, 85mm ID X 206mm Inside Depth, 107mm OD X 231mm Height
SDP
Catalog No. 50872091
Encompass_Preferred
Flask, Dewar, 1000mL, Narrow Mouth, Stainless Steel, Carrying Handle, 85mm ID X 206mm Inside Depth, 107mm OD X 231mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel. Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage and transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200°C and has a carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.
Catalog No. 50-872-091 Supplier Chemglass Life Sciences Supplier No. CG1599B02
Flask, Dewar, 1000mL, Narrow Mouth, Stainless Steel, Carrying Handle, 85mm ID X 206mm Inside Depth, 107mm OD X 231mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel. Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage and transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200°C and has a carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.