Learn More
Chemglass Life Sciences Flask, Dewar, 1000mL, Narrow Mouth, Stainless Steel, Side Grip and Carrying Handle, 85mm ID X 206mm Inside Depth, 107mm OD X 231mm Height
SDP
Catalog No. 501531174
Encompass_Preferred
Flask, Dewar, 1000mL, Narrow Mouth, Stainless Steel , Side Grip and Carrying Handle, 85mm ID X 206mm Inside Depth, 107mm OD X 231mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel . Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage & transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200C and has a side grip & carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.
Catalog No. 50-153-1174 Supplier Chemglass Life Sciences Supplier No. CG1599A02
Flask, Dewar, 1000mL, Narrow Mouth, Stainless Steel , Side Grip and Carrying Handle, 85mm ID X 206mm Inside Depth, 107mm OD X 231mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel . Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage & transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200C and has a side grip & carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.