Learn More
Chemglass Life Sciences Flask, Dewar, 2L, Stainless Steel, Carrying Handle, 100mm ID X 285mm Inside Depth, 122mm OD X 312mm Height
SDP
Catalog No. 501531180
Encompass_Preferred
Flask, Dewar, 2L, Stainless Steel , Carrying Handle, 100mm ID X 285mm Inside Depth, 122mm OD X 312mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel . Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage and transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks ahve a temperature range of -200 to +200C and has a carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.
Catalog No. 50-153-1180 Supplier Chemglass Life Sciences Supplier No. CG1599B04
Flask, Dewar, 2L, Stainless Steel , Carrying Handle, 100mm ID X 285mm Inside Depth, 122mm OD X 312mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel . Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage and transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks ahve a temperature range of -200 to +200C and has a carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.