Learn More
Chemglass Life Sciences Flask, Dewar, 2L, Stainless Steel, Side Grip and Carrying Handle, 100mm ID X 285mm Inside Depth, 122mm OD X 312mm Height
SDP
Catalog No. 50872087
Encompass_Preferred
Flask, Dewar, 2L, Stainless Steel, Side Grip and Carrying Handle, 100mm ID X 285mm Inside Depth, 122mm OD X 312mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel. Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage & transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200°C and has a side grip & carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.
Catalog No. 50-872-087 Supplier Chemglass Life Sciences Supplier No. CG1599A04
Flask, Dewar, 2L, Stainless Steel, Side Grip and Carrying Handle, 100mm ID X 285mm Inside Depth, 122mm OD X 312mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel. Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage & transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200°C and has a side grip & carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug.