Learn More
Chemglass Life Sciences Flask, Dewar, 500mL Stainless Steel, Side Grip Handle Only, 65mm ID X 180mm Inside Depth, 87mm OD X 203mm Height
SDP
Catalog No. 501531173
Encompass_Preferred
Flask, Dewar, 500mL Stainless Steel , Side Grip Handle Only, 65mm ID X 180mm Inside Depth, 87mm OD X 203mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel . Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage & transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200C and has a side grip & carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug. PLEASE NOTE: CG-1599-A-01 HAS A SIDE GRIP HANDLE, BUT DOES NOT HAVE THE CARRYING HANDLE.
Catalog No. 50-153-1173 Supplier Chemglass Life Sciences Supplier No. CG1599A01
Flask, Dewar, 500mL Stainless Steel , Side Grip Handle Only, 65mm ID X 180mm Inside Depth, 87mm OD X 203mm Height. Unbreakable Dewar flask is made entirely of Stainless Steel . Dewar flask has many flexible laboratory applications including cooling of small samples and as an insulating flask. Furthermore, it can be used for storage & transport of liquid nitrogen LN2, dry ice CO2, and other coolants. Dewar flasks have a temperature range of -200 to +200C and has a side grip & carrying handle for easy handling. See CG-1599-S for optional Loose Lid Insulating Plug. PLEASE NOTE: CG-1599-A-01 HAS A SIDE GRIP HANDLE, BUT DOES NOT HAVE THE CARRYING HANDLE.