Learn More
Hach Company Flask-Digesdahl
Supplier:  Hach Company 2312542
Encompass
Specially designed flat-bottom glass flask for use with the Hach Digesdahl Digestion Apparatus Each flask is calibrated individually with a 100 mL (to contain) mark
Catalog No. 50-002-56973