Learn More
Hach Company Flask, Erlenmeyer, Polymethylpentene Capacity 500 mL, 4/pk
Catalog No. 501995317 Shop All Hach Company Products
Encompass
Polymethylpentene flask with polypropylene screwcap features clarity, chemical resistance, strength, and autoclavability (Do not engage threads during cycle). Useful for titrations, preparation and storage of media.
Catalog No. 50-199-5317 Supplier Hach Company Supplier No. 2089879
Polymethylpentene flask with polypropylene screwcap features clarity, chemical resistance, strength, and autoclavability (Do not engage threads during cycle). Useful for titrations, preparation and storage of media.