Learn More
Hach Company Glass Erlenmeyer Flask, 500 mL, with Screw Cap, 6 pack
Supplier:  Hach Company 2089679
Encompass
Graduated glass flask with plastic screw cap. Excellent choice for media preparation, mixing solutions, and culturing. Inert rubber liner cemented into cap and completely autoclavable. Durable graduations indicate approximate volume.
Catalog No. 50-199-5309