Learn More
Chemglass Life Sciences Chemglass Erlenmeyer flask, polycarbonate, 1,000 mL, baffled, vent filter cap, sterile (CGN-2091-1000)
SDP
Supplier:  Chemglass Life Sciences CGN20911000
Encompass_Preferred
Sterile 1,000 mL polycarbonate Erlenmeyer flask designed for cell culture and bioprocess applications. The baffled flat base improves mixing and aeration, while the vent filter cap provides gas exchange without contamination. Molded graduations enable convenient volume readback, and flasks are individually bagged and E-beam sterilized for immediate use.
  • Baffled flat bottom improves mixing and oxygen transfer.
  • Vented cap with 0.2 μm hydrophobic filter for sterile gas exchange.
  • Made from polycarbonate for shatter resistance and lightweight handling.
  • Molded graduations for accurate volume readback.
  • Individually bagged and E-beam sterilized; non-pyrogenic and DNase/RNase-free.
  • Packaged as case of 6 (CS6).
  • Dimensions: height 213.3 mm, neck diameter 39 mm, bottom diameter 127 mm.
Catalog No. 50-273-3482