Learn More
Chemglass Life Sciences Erlenmeyer Flask, 1,000 mL, Polycarbonate, Plain, Sterile, 38-430 Thread with Blue Duo Cap, Case of 24
SDP
Catalog No. 502733486
Encompass_Preferred
Sterile, non-pyrogenic polycarbonate Erlenmeyer flask suitable for cell culture and general laboratory mixing, shaking, and storage. The DuoCap can be used as a vented filter cap or converted to a solid top for mixing and storage.
  • Sterile, non-pyrogenic polycarbonate construction.
  • 1,000 mL capacity with molded graduations for accurate volume indication.
  • 38-430 thread with DuoCap allowing vented or solid-top use.
  • Vented option uses a 0.22 μm PTFE membrane for sterile air exchange.
  • Graduations molded for accuracy and repeated use.
  • EZTear packaging for easy access.
  • Packaged in a case quantity of 24 for bulk laboratory use.
Catalog No. 50-273-3486 Supplier Chemglass Life Sciences Supplier No. CGN20901000
Sterile, non-pyrogenic polycarbonate Erlenmeyer flask suitable for cell culture and general laboratory mixing, shaking, and storage. The DuoCap can be used as a vented filter cap or converted to a solid top for mixing and storage.
  • Sterile, non-pyrogenic polycarbonate construction.
  • 1,000 mL capacity with molded graduations for accurate volume indication.
  • 38-430 thread with DuoCap allowing vented or solid-top use.
  • Vented option uses a 0.22 μm PTFE membrane for sterile air exchange.
  • Graduations molded for accuracy and repeated use.
  • EZTear packaging for easy access.
  • Packaged in a case quantity of 24 for bulk laboratory use.