Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, Polycarbonate, 1,000 mL, Flat Bottom, Vent Filter Cap, Sterile (Nest)
SDP
Catalog No. 502733476
Encompass_Preferred
A 1,000 mL sterile polycarbonate Erlenmeyer flask suitable for laboratory mixing, culture, and shaking applications. The flat-bottom design provides stable placement, while the vent filter cap allows sterile gas exchange and helps reduce contamination risk.
  • Polycarbonate construction for durability and clarity.
  • 1,000 mL nominal capacity.
  • Flat bottom for stable placement on surfaces.
  • Vent filter cap enables sterile aeration.
  • Nest packaging for sterile, space-efficient storage.
Catalog No. 50-273-3476 Supplier Chemglass Life Sciences Supplier No. CGN20941000
A 1,000 mL sterile polycarbonate Erlenmeyer flask suitable for laboratory mixing, culture, and shaking applications. The flat-bottom design provides stable placement, while the vent filter cap allows sterile gas exchange and helps reduce contamination risk.
  • Polycarbonate construction for durability and clarity.
  • 1,000 mL nominal capacity.
  • Flat bottom for stable placement on surfaces.
  • Vent filter cap enables sterile aeration.
  • Nest packaging for sterile, space-efficient storage.