Learn More
Chemglass Life Sciences Flask, Erlenemeyer, 10mL, GPI 20-400 Thread
SDP
Catalog No. 501531781
Encompass_Preferred
Flask, Erlenemeyer, 10mL, GPI 20-400 Thread. Erlenmeyer flask with a GPI 20-400 thread. For use with Mininert valves. Ideal for extraction of Trihalomethanes (EPA Method 501.2).
Catalog No. 50-153-1781 Supplier Chemglass Life Sciences Supplier No. CG4914A01
Flask, Erlenemeyer, 10mL, GPI 20-400 Thread. Erlenmeyer flask with a GPI 20-400 thread. For use with Mininert valves. Ideal for extraction of Trihalomethanes (EPA Method 501.2).