Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 125mL, Plain, Sterile, Polycarbonate, 38-430 Thread, with Blue Duo Cap
SDP
Catalog No. 501532279
Encompass_Preferred
Flask, Erlenmeyer, 125mL, Plain, Sterile, Polycarbonate, 38-430 Thread, with Blue Duo Cap. Erlenmeyer flasks are sterile, non-pyrogenic and manufactured with polycarbonate resin. Flasks are supplied with exclusive DuoCap designed for both aerobic and anaerobic applications. For aerobic applications the cap utilizes a 0.22m pore size PTFE membrane for sterile air exchange. For anaerobic applications the cap can easily be converted to a solid top design. Solid top caps are also ideal for applications requiring mixing, shaking and storage of biological fluids. Graduations are molded for accuracy.EZTear packaging allows for easy access.
Catalog No. 50-153-2279 Supplier Chemglass Life Sciences Supplier No. CLS2090125B
Flask, Erlenmeyer, 125mL, Plain, Sterile, Polycarbonate, 38-430 Thread, with Blue Duo Cap. Erlenmeyer flasks are sterile, non-pyrogenic and manufactured with polycarbonate resin. Flasks are supplied with exclusive DuoCap designed for both aerobic and anaerobic applications. For aerobic applications the cap utilizes a 0.22m pore size PTFE membrane for sterile air exchange. For anaerobic applications the cap can easily be converted to a solid top design. Solid top caps are also ideal for applications requiring mixing, shaking and storage of biological fluids. Graduations are molded for accuracy.EZTear packaging allows for easy access.