Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 125mL, GL-32 Thread
SDP
Catalog No. 501311071
Encompass_Preferred
Flask, Erlenmeyer, 125mL, GL-32 Thread. Flask has a GL screw thread top and is supplied complete with a polypropylene liner less closure and pouring ring. Graduated to approximate volumes.
Catalog No. 50-131-1071 Supplier Chemglass Life Sciences Supplier No. CG154419
Flask, Erlenmeyer, 125mL, GL-32 Thread. Flask has a GL screw thread top and is supplied complete with a polypropylene liner less closure and pouring ring. Graduated to approximate volumes.