Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, 125 mL, Polycarbonate, Flat Bottom, Seal Cap, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2090125
Encompass_Preferred
125 mL polycarbonate Erlenmeyer flask with a flat bottom and seal cap, supplied sterile and non-pyrogenic. The flask is DNase/RNase-free, individually packaged in sterile bags, and suitable for cell culture and general laboratory liquid handling.
  • 125 mL capacity.
  • Manufactured from polycarbonate.
  • Flat bottom with a conical body.
  • Seal cap included.
  • Sterile. E-beam sterilized (SAL = 10).
  • Non-pyrogenic and DNase/RNase-free.
  • Individually packaged in sterile bags.
  • Dimensions: height 106.8 mm; neck dia. 34 mm; bottom dia. 66 mm.
Catalog No. 50-273-3485