Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, Polycarbonate, 125 mL, Flat Bottom, Vent Filter Cap, Sterile (CGN-2094-125)
SDP
Catalog No. 502733475
Encompass_Preferred
Polycarbonate Erlenmeyer flask with 125 mL capacity, conical shape and flat bottom. Supplied sterile with a vent filter cap to allow gas exchange while minimizing contamination. Lightweight and shatter-resistant, suitable for shaker-based culture and general laboratory use.
  • Conical, flat-bottom design for stable mixing
  • Polycarbonate construction: lightweight and shatter-resistant
  • Vent filter cap permits gas exchange while reducing contamination
  • Supplied sterile for single-use applications
  • Suitable for shaker-based cell culture and general laboratory use
Catalog No. 50-273-3475 Supplier Chemglass Life Sciences Supplier No. CGN2094125
Polycarbonate Erlenmeyer flask with 125 mL capacity, conical shape and flat bottom. Supplied sterile with a vent filter cap to allow gas exchange while minimizing contamination. Lightweight and shatter-resistant, suitable for shaker-based culture and general laboratory use.
  • Conical, flat-bottom design for stable mixing
  • Polycarbonate construction: lightweight and shatter-resistant
  • Vent filter cap permits gas exchange while reducing contamination
  • Supplied sterile for single-use applications
  • Suitable for shaker-based cell culture and general laboratory use