Learn More
Chemglass Life Sciences Chemglass Erlenmeyer flask, 125 mL, polycarbonate, quadruple-baffled, sterile, seal cap
SDP
Catalog No. 502733479
Encompass_Preferred
Sterile, non-pyrogenic 125 mL Erlenmeyer flask made from polycarbonate resin for mixing, shaking, and storage of biological fluids. The quadruple-baffled bottom improves aeration and mixing, and molded graduations support accurate measurements. Supplied individually in sterile packaging.
  • Capacity: 125 mL.
  • Material: Polycarbonate resin.
  • Sterilization: E-beam (SAL = 10).
  • Pyrogen status: Non-pyrogenic; DNase/RNase-free.
  • Bottom: Quadruple baffled to improve mixing and aeration.
  • Cap: Seal cap suitable for anaerobic applications and storage.
  • Graduations: Molded graduations for accurate volume reading.
  • Packaging: Individually packaged in sterile bags.
Catalog No. 50-273-3479 Supplier Chemglass Life Sciences Supplier No. CGN2093125
Sterile, non-pyrogenic 125 mL Erlenmeyer flask made from polycarbonate resin for mixing, shaking, and storage of biological fluids. The quadruple-baffled bottom improves aeration and mixing, and molded graduations support accurate measurements. Supplied individually in sterile packaging.
  • Capacity: 125 mL.
  • Material: Polycarbonate resin.
  • Sterilization: E-beam (SAL = 10).
  • Pyrogen status: Non-pyrogenic; DNase/RNase-free.
  • Bottom: Quadruple baffled to improve mixing and aeration.
  • Cap: Seal cap suitable for anaerobic applications and storage.
  • Graduations: Molded graduations for accurate volume reading.
  • Packaging: Individually packaged in sterile bags.