Learn More
Chemglass Life Sciences Flask, Shake, 2000mL, 6 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 170 x 270
SDP
Catalog No. 501532277
Encompass_Preferred
Flask, Shake, 2000mL, 6 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 170 x 270. Flask has 6 baffles, 3 baffles on the lower outside edge of the flask and 3 baffles on the bottom surface of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. Gray polypropylene vented cap has a 0.22m (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination. They have a liner-less plug seal design for efficient sealing. They are autoclavable at 121C. Since these caps are vented, the flasks can be autoclaved with the cap attached. Please Note: Glass Shake and Fernbach flasks are not designed for use with a SOLID screw cap for culture of anaerobic microorganisms. (Pressure resistant bottles, flasks or tubes are recommended).
Catalog No. 50-153-2277 Supplier Chemglass Life Sciences Supplier No. CLS205210
Flask, Shake, 2000mL, 6 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 170 x 270. Flask has 6 baffles, 3 baffles on the lower outside edge of the flask and 3 baffles on the bottom surface of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. Gray polypropylene vented cap has a 0.22m (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination. They have a liner-less plug seal design for efficient sealing. They are autoclavable at 121C. Since these caps are vented, the flasks can be autoclaved with the cap attached. Please Note: Glass Shake and Fernbach flasks are not designed for use with a SOLID screw cap for culture of anaerobic microorganisms. (Pressure resistant bottles, flasks or tubes are recommended).