Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, Polycarbonate, 2,000 mL, Sterile, Seal Cap, Flat Bottom
SDP
Catalog No. 502733472
Encompass_Preferred
Sterile 2,000 mL polycarbonate Erlenmeyer flask designed for mixing, shaking, and storage of biological fluids. Supplied with a seal cap and molded graduations, the flask is sterilized by E-beam and packaged individually for aseptic workflows.
  • 2,000 mL capacity with molded graduations for accurate measurement.
  • Polycarbonate construction provides clarity and impact resistance.
  • Supplied with a seal cap suitable for anaerobic applications.
  • Sterilized by E-beam (SAL=10); non-pyrogenic and DNase/RNase free.
  • Flat bottom, conical shape for stable mixing and uniform heating.
  • Individually packaged in sterile bags for aseptic workflows.
  • Dimensions: height 285 mm; neck outside diameter 47 mm; bottom diameter 162 mm.
Catalog No. 50-273-3472 Supplier Chemglass Life Sciences Supplier No. CGN20952000
Sterile 2,000 mL polycarbonate Erlenmeyer flask designed for mixing, shaking, and storage of biological fluids. Supplied with a seal cap and molded graduations, the flask is sterilized by E-beam and packaged individually for aseptic workflows.
  • 2,000 mL capacity with molded graduations for accurate measurement.
  • Polycarbonate construction provides clarity and impact resistance.
  • Supplied with a seal cap suitable for anaerobic applications.
  • Sterilized by E-beam (SAL=10); non-pyrogenic and DNase/RNase free.
  • Flat bottom, conical shape for stable mixing and uniform heating.
  • Individually packaged in sterile bags for aseptic workflows.
  • Dimensions: height 285 mm; neck outside diameter 47 mm; bottom diameter 162 mm.