Learn More
Chemglass Life Sciences Chemglass Erlenmeyer Flask, Polycarbonate, 2,000 mL, Sterile, Vent Filter Cap (CGN-2096-2000)
SDP
Catalog No. 502733470
Encompass_Preferred
Sterile 2,000 mL polycarbonate Erlenmeyer flask with a flat bottom and conical body, supplied with a vent filter cap configured for gas exchange while minimizing contamination.
  • Sterilized by E-beam, SAL=10.
  • Made from durable, transparent polycarbonate.
  • Vent filter cap with 0.2 μm hydrophobic membrane for gas exchange.
  • Non-pyrogenic and DNase/RNase free.
  • Molded graduations for accurate volume reading.
  • Flat bottom for stable placement and conical shape for efficient mixing.
  • Individually packaged in sterile bags; case pack of 6.
Catalog No. 50-273-3470 Supplier Chemglass Life Sciences Supplier No. CGN20962000
Sterile 2,000 mL polycarbonate Erlenmeyer flask with a flat bottom and conical body, supplied with a vent filter cap configured for gas exchange while minimizing contamination.
  • Sterilized by E-beam, SAL=10.
  • Made from durable, transparent polycarbonate.
  • Vent filter cap with 0.2 μm hydrophobic membrane for gas exchange.
  • Non-pyrogenic and DNase/RNase free.
  • Molded graduations for accurate volume reading.
  • Flat bottom for stable placement and conical shape for efficient mixing.
  • Individually packaged in sterile bags; case pack of 6.