Learn More
Chemglass Life Sciences Erlenmeyer Flask, 250 mL, Polycarbonate, Flat Bottom, Sterile, Seal Cap
SDP
Catalog No. 502733484
Encompass_Preferred
Sterile 250 mL polycarbonate Erlenmeyer flask designed for mixing, shaking, and storage of biological fluids. Molded graduations provide accurate measurements; flasks are non-pyrogenic and DNase/RNase-free. Sterilized by E-beam (SAL = 10) and supplied in sterile packaging.
  • Capacity: 250 mL.
  • Material: Polycarbonate.
  • Flat bottom for stable placement.
  • Seal cap suitable for anaerobic applications and mixing.
  • Sterile, E-beam sterilized with SAL = 10.
  • Non-pyrogenic and DNase/RNase-free.
  • Molded graduations for accurate measurement.
  • Packaged individually; 12 per carton.
Catalog No. 50-273-3484 Supplier Chemglass Life Sciences Supplier No. CGN2090250
Sterile 250 mL polycarbonate Erlenmeyer flask designed for mixing, shaking, and storage of biological fluids. Molded graduations provide accurate measurements; flasks are non-pyrogenic and DNase/RNase-free. Sterilized by E-beam (SAL = 10) and supplied in sterile packaging.
  • Capacity: 250 mL.
  • Material: Polycarbonate.
  • Flat bottom for stable placement.
  • Seal cap suitable for anaerobic applications and mixing.
  • Sterile, E-beam sterilized with SAL = 10.
  • Non-pyrogenic and DNase/RNase-free.
  • Molded graduations for accurate measurement.
  • Packaged individually; 12 per carton.