Learn More
Chemglass Life Sciences Chemglass 250 mL Polycarbonate Erlenmeyer Flask, Flat Bottom, Vent Filter Cap, Sterile, Individually Bagged
SDP
Catalog No. 502733474
Encompass_Preferred
250 mL polycarbonate Erlenmeyer flask designed for cell culture and general laboratory mixing. The flask provides a sterile, vented container with molded graduations and a stable flat bottom; supplied individually bagged for aseptic handling.
  • 250 mL capacity.
  • Polycarbonate construction.
  • Flat bottom, conical body.
  • Vent filter cap for filtered gas exchange.
  • Individually bagged sterile packaging.
  • Molded graduations for accurate measurements.
  • Non-pyrogenic and DNase/RNase free.
  • E-beam sterilized (SAL=10).
  • Approx. dimensions: height 137.8 mm; neck diameter 34 mm; base diameter 83 mm.
Catalog No. 50-273-3474 Supplier Chemglass Life Sciences Supplier No. CGN2094250
250 mL polycarbonate Erlenmeyer flask designed for cell culture and general laboratory mixing. The flask provides a sterile, vented container with molded graduations and a stable flat bottom; supplied individually bagged for aseptic handling.
  • 250 mL capacity.
  • Polycarbonate construction.
  • Flat bottom, conical body.
  • Vent filter cap for filtered gas exchange.
  • Individually bagged sterile packaging.
  • Molded graduations for accurate measurements.
  • Non-pyrogenic and DNase/RNase free.
  • E-beam sterilized (SAL=10).
  • Approx. dimensions: height 137.8 mm; neck diameter 34 mm; base diameter 83 mm.