Learn More
Chemglass Life Sciences Erlenmeyer Flask, Flat-Bottom, 3,000 mL Polycarbonate, Sterile, Vent Filter Cap
SDP
Catalog No. 502733469
Encompass_Preferred
Sterile 3,000 mL polycarbonate Erlenmeyer flask with a flat bottom and vent filter cap, suitable for mixing, culturing, and general laboratory applications. Molded graduations and clear material allow easy observation and measurement.
  • Polycarbonate construction for high transparency and durability.
  • 3,000 mL capacity with molded graduation marks for accurate volume estimation.
  • Flat bottom for stable placement on lab surfaces.
  • Vent filter cap with 0.2 μm hydrophobic filter to allow gas exchange while limiting contamination.
  • Sterilized by e-beam with SAL = 10 and individually packaged in sterile bags.
  • Non-pyrogenic and DNase/RNase free.
  • Dimensions: height 253 mm; bottom diameter 230 mm; neck outside diameter 62 mm.
Catalog No. 50-273-3469 Supplier Chemglass Life Sciences Supplier No. CGN20963000
Sterile 3,000 mL polycarbonate Erlenmeyer flask with a flat bottom and vent filter cap, suitable for mixing, culturing, and general laboratory applications. Molded graduations and clear material allow easy observation and measurement.
  • Polycarbonate construction for high transparency and durability.
  • 3,000 mL capacity with molded graduation marks for accurate volume estimation.
  • Flat bottom for stable placement on lab surfaces.
  • Vent filter cap with 0.2 μm hydrophobic filter to allow gas exchange while limiting contamination.
  • Sterilized by e-beam with SAL = 10 and individually packaged in sterile bags.
  • Non-pyrogenic and DNase/RNase free.
  • Dimensions: height 253 mm; bottom diameter 230 mm; neck outside diameter 62 mm.