Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 4000mL, GL-45 Thread
SDP
Catalog No. 501531165
Encompass_Preferred
Flask, Erlenmeyer, 4000mL, GL-45 Thread. Flask has a GL screw thread top and is supplied complete with a polypropylene liner less closure and pouring ring. Graduated to approximate volumes.
Catalog No. 50-153-1165 Supplier Chemglass Life Sciences Supplier No. CG154424
Flask, Erlenmeyer, 4000mL, GL-45 Thread. Flask has a GL screw thread top and is supplied complete with a polypropylene liner less closure and pouring ring. Graduated to approximate volumes.