Learn More
Chemglass Life Sciences Nest 500 mL Polycarbonate Erlenmeyer Flask, Conical Flat Bottom with 0.2 μm Vent Filter Cap, Sterile
SDP
Catalog No. 502733473
Encompass_Preferred
This sterile 500 mL polycarbonate Erlenmeyer flask features a conical shape and flat bottom, and is supplied with a 0.2 μm vent filter cap for contamination-free gas exchange. Manufactured from polycarbonate for clarity and durability, the flask is non-pyrogenic, DNase/RNase-free, and sterilized by E-beam (SAL 10). Individually bagged for aseptic use.
  • 500 mL capacity for media preparation and culture.
  • Polycarbonate construction provides optical clarity and impact resistance.
  • Conical shape with flat bottom suitable for shaking and incubating.
  • Vent filter cap with 0.2 μm hydrophobic filter enables gas exchange without contamination.
  • Sterilized by E-beam (SAL 10) and individually bagged.
  • Non-pyrogenic and DNase/RNase-free for molecular biology applications.
  • Molded graduations provide approximate volume measurement.
Catalog No. 50-273-3473 Supplier Chemglass Life Sciences Supplier No. CGN2094500
This sterile 500 mL polycarbonate Erlenmeyer flask features a conical shape and flat bottom, and is supplied with a 0.2 μm vent filter cap for contamination-free gas exchange. Manufactured from polycarbonate for clarity and durability, the flask is non-pyrogenic, DNase/RNase-free, and sterilized by E-beam (SAL 10). Individually bagged for aseptic use.
  • 500 mL capacity for media preparation and culture.
  • Polycarbonate construction provides optical clarity and impact resistance.
  • Conical shape with flat bottom suitable for shaking and incubating.
  • Vent filter cap with 0.2 μm hydrophobic filter enables gas exchange without contamination.
  • Sterilized by E-beam (SAL 10) and individually bagged.
  • Non-pyrogenic and DNase/RNase-free for molecular biology applications.
  • Molded graduations provide approximate volume measurement.