Learn More
Chemglass Life Sciences Chemglass Erlenmeyer flask, 500 mL, polycarbonate, baffled bottom, sterile, with seal cap (nest packaged)
SDP
Catalog No. 502733477
Encompass_Preferred
This 500 mL baffled Erlenmeyer flask is made from polycarbonate and supplied sterile in nest packaging for laboratory culture, mixing, shaking, and storage applications. The flask includes molded graduations for accurate volume measurement and is sterilized by E-beam to SAL=10.
  • 500 mL capacity.
  • Polycarbonate construction for strength and clarity.
  • Quadruple baffled bottom to improve aeration during shaking.
  • Seal cap provided for anaerobic applications.
  • Solid caps suitable for mixing, shaking, and storage of biological fluids.
  • Molded graduations for accurate measurements.
  • Sterilized by E-beam; SAL=10.
  • Non-pyrogenic and DNase/RNase-free.
  • Individually packaged in sterile bags (nest packaging).
Catalog No. 50-273-3477 Supplier Chemglass Life Sciences Supplier No. CGN2093500
This 500 mL baffled Erlenmeyer flask is made from polycarbonate and supplied sterile in nest packaging for laboratory culture, mixing, shaking, and storage applications. The flask includes molded graduations for accurate volume measurement and is sterilized by E-beam to SAL=10.
  • 500 mL capacity.
  • Polycarbonate construction for strength and clarity.
  • Quadruple baffled bottom to improve aeration during shaking.
  • Seal cap provided for anaerobic applications.
  • Solid caps suitable for mixing, shaking, and storage of biological fluids.
  • Molded graduations for accurate measurements.
  • Sterilized by E-beam; SAL=10.
  • Non-pyrogenic and DNase/RNase-free.
  • Individually packaged in sterile bags (nest packaging).