Learn More
MTC Bio hybex™ Glass Erlenmeyer Flask, 50ml, 12/pk
Catalog No. NC1859324
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Dishes, plates and flasks, Flasks, Erlenmeyer flasks, hybex Glass Erlenmeyer Flasks, 50ml. Made from high quality, borosilicate 3.3 glass, low coefficient of thermal expansion, high temperature resistance, permanent white enamel graduations and labeling area. Ideal for shaking samples and media preparation. Pack of 12.
Catalog No. NC1859324 Supplier MTC Bio Supplier No. TB311050
This item has a minimum qty of 3 per supplier requirements.
Dishes, plates and flasks, Flasks, Erlenmeyer flasks, hybex Glass Erlenmeyer Flasks, 50ml. Made from high quality, borosilicate 3.3 glass, low coefficient of thermal expansion, high temperature resistance, permanent white enamel graduations and labeling area. Ideal for shaking samples and media preparation. Pack of 12.