Learn More
Chemglass Life Sciences Erlenmeyer flask, 5 L polycarbonate, baffled bottom, sterile - vent or seal cap
SDP
Supplier:  Chemglass Life Sciences CGN21665LSC
Encompass_Preferred
High-efficiency 5 L Erlenmeyer flask made from medical-grade polycarbonate for shaken culture and mixing applications. The baffled bottom improves aeration and mixing; the flask is supplied sterile and meets USP Class VI (ISO 10993) biocompatibility standards. Available with vent filter or seal cap options.
  • Polycarbonate construction with high transparency and impact resistance.
  • Baffled bottom for improved aeration and mixing.
  • Sterile, e-beam sterilized and non-pyrogenic.
  • Meets USP Class VI / ISO 10993 biocompatibility requirements.
  • Clear graduation scale for easy volume observation.
  • Recommended working volume 30-40% of total volume.
  • Withstands temperatures up to 121 °C.
  • Packaged in sterile nests, 4 per case.
  • Vent cap includes a 0.22 μm breathable filter for gas exchange when selected.
Catalog No. 50-273-3441