Learn More
Hach Company Flask, Erlenmeyer, Polypropylene, 125 mL, pk/12
Supplier:  Hach Company 108273
Encompass
Polypropylene flask for general laboratory use. Rugged, excellent chemical resistance, and autoclavable. Stopper not included.
Catalog No. 50-199-4541