Learn More
Hach Company Flask, Erlenmeyer, Polypropylene, 250 mL
Supplier:  Hach Company 108246
Encompass
Polypropylene flask for general laboratory use. Rugged, excellent chemical resistance, and autoclavable. Stopper not included.
Catalog No. 50-199-4540