Learn More
Hach Company Erlenmeyer Glass Flask, 50 mL, with Screw Cap
Supplier: Hach Company 2089641
Graduated glass flask with plastic screw cap. Excellent choice for media preparation, mixing solutions, and culturing. Inert rubber liner cemented into cap and completely autoclavable. Durable graduations indicate approximate volume.