Learn More
Chemglass Life Sciences 500mL Erlenmeyer Flask, 24/40 Outer Joint, Graduated
SDP
Catalog No. CG154209
Encompass_Preferred
500mL Erlenmeyer Flask, 24/40 Outer Joint, Graduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.
Catalog No. CG154209 Supplier Chemglass Life Sciences Supplier No. CG154209
500mL Erlenmeyer Flask, 24/40 Outer Joint, Graduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.