Learn More
Chemglass Life Sciences 1000mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask
SDP
Catalog No. 50873109
Encompass_Preferred
1000mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.
Catalog No. 50-873-109 Supplier Chemglass Life Sciences Supplier No. CG1512P09
1000mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.