Learn More
Chemglass Life Sciences 500mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask
SDP
Supplier:  Chemglass Life Sciences CG1512P07
Encompass_Preferred
500mL Single Neck Evaporating Flask, 24/40 Outer Joint, Plastic Coated Recovery Flask. Similar to CG-1512 recovery flasks, but plastic coated to prevent injury from broken glass and spillage of hazardous materials.
Catalog No. 50-873-108